missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Rack Slanter
Tilts test tube rack to 25°, 35° or 45°
Supplier: Bel-Art™ F188900100
Description
- Increases the surface area of agar inside test tubes
- Decreases hand strain when working with pipetters by adjusting the rack angle
- Fits racks up to 11.56L x 9.56W x 3.25 in. H (29.3 x 24.2 x 8.3 cm)
- Base legs have predetermined slots that stand slides in and out of to easily change angles
- Rigid polypropylene construction
- Steam autoclavable at 121°C (250°F)
- Also for use with the Mini-Magnifier (Mfr. No. 378650000) to aid in the viewing of Petri dishes
Specifications
| Test Tube Slant Rack | |
| Autoclavable | |
| Test Tubes | |
| 3 | |
| None | |
| Non-sterile |
| 1 x 1 | |
| Translucent | |
| Polypropylene | |
| Rectangular | |
| No |