Learn More
Bel-Art™ Pipet Vacuum Aspirator Collection System
Catalog No. 13717261
Change view
Click to view available options
Type:
Pipet Vacuum Aspirator Collection System
Replacement Hydrophobic Filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13717261 Replacement Hydrophobic Filter
13717260 Pipet Vacuum Aspirator Collection System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13717261 Supplier Bel-Art™ Supplier No. F199170155
Get Up to $500 in Products with Purchase
View all promotions
Performs vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes.
  • Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
  • Maintains low noise level (76 db at 1 meter) and minimal vibration
  • Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
  • Chemical and corrosion resistant polypropylene collection bottle holds 3800 mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
  • A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
  • Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
For Use With Pipettes
Material Plastic
Type Replacement Hydrophobic Filter
Volume (Metric) 3800 mL
For Use With (Equipment) Bel-Art Vacuum Aspirator Collection System