missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ OdoClave™ Deodorant Autoclave Pads

Catalog No. 13717292
Change view
Click to view available options
Scent:
Cherry
Clove
Lemon
Pine
Triple-strength Clove
Color:
Amber
Brown
Green
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent Color
13717292 Lemon Amber
1448831 Triple-strength Clove Yellow
1448830 Clove Brown
1448832 Cherry Red
1448833 Pine Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13717292 Supplier Bel-Art™ Supplier No. H131980003
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Neutralize autoclave odors with a pleasant fragrance

  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • 32mm (1.75 in.) square
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads

Order Info

Choose 1.25 in. (32mm) square regular-strength or 2.25 in. (5.7cm) square super-strength pad.

Specifications

Product Type Autoclave Capsule
Color-coded No
Form Liquid
Scent Lemon
Sterility Non-sterile
Autoclavable Autoclavable
Color Amber
Format Pad
For Use With (Application) Autoclave