missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Manomark™ Pens

Catalog No. 133851
Change view
Click to view available options
Color:
Assorted
Black
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
133851 Black
13385 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 133851 Supplier Bel-Art™ Supplier No. F133770001
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth.

  • Solvent-based ink is hydrocarbon, water and temperature resistant 0°C to 155°C (32°F to 310°F)
  • Marks can be erased with alcohol
  • Unique seal prevents drying out if pen is left uncapped

Specifications

Tip Style Felt Tip
Product Type Pen
Color Black
For Use With (Application) Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth
Temperature (English) Range 32°F to 310°F
Temperature (Metric) Range 0°C to 155°C
Quantity 12 Pk.