missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Magnetic Bead Separation Racks

Catalog No. p-5159674
Change view
Click to view available options
Holds:
Microplates (Standard Size Flat)
PCR Tube Plate (96-well)
Tubes (2 mL)
Tubes (5 or 15 mL)
Tubes (50 mL)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
13717262 Tubes (50 mL)
13717263 Tubes (5 or 15 mL)
13717265 PCR Tube Plate (96-well)
13717266 Microplates (Standard Size Flat)
13717264 Tubes (2 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13717262 Supplier Bel Art™ Supplier No. F199000000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays.

  • Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves of 1.5 to 50 mL models allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
  • 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes
  • Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96 etc.
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Tubes (50 mL)
Material Neodymium Magnets, Acrylic
Positions 3
Shape Cylindrical
Stackable No
Sterility Non-sterile
For Use With (Application) General Laboratories, Immunology, Proteomics, Genomics, Separation Methods
Product Type Magnetic Bead Separation Rack
Shelves None
Height (Metric) 76 mm
Array 1 x 3
Width (Metric) 44 mm
Show More Show Less