Learn More
Bel Art™ Lab Companion Cabinet Style Vacuum Desiccator, Amber
UV blocking amber desiccator provides vacuum sustainment in a space maximizing cube.
$2,698.76 - $5,435.98
Specifications
Color Amber
Material Polycarbonate
Product Type Vacuum Desiccator
Door Type Hinged
Style Cabinet
View More Specs
Includes: Desiccant tray (desiccants sold separately)
Products 4
Catalog Number Mfr. No. Volume (Metric) No. of Shelves Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) No. of Shelves Price Quantity & Availability  
08648110
View Documents Promotion Details
Bel Art™
F424004101
11 L 2/4
Each for $2,698.76
Only null left
08648111
View Documents Promotion Details
Bel Art™
F424004111
35 L 3/5
Each for $4,245.53
Only null left
08648112
View Documents Promotion Details
Bel Art™
F424004121
23 L 2/4
Each for $3,333.29
Only null left
08648113
View Documents Promotion Details
Bel Art™
F424004131
45 L 3/6
Each for $5,435.98
Only null left
Description
Description
  • High-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind.
  • Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods.
  • Viton™ 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange.
  • Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents.
  • Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
  • Use with any vacuum with free air capacity of 1.5 cubic feet per minute (CFM) ft or greater.
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit.
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage.
  • Locking ring secures top and bottom halves when vacuum not in use; provides additional convenience and safety when moving the unit.
  • Six models to choose from; 6, 10, and 20 liter internal volume in clear or UV blocking amber.
Specifications
Specifications
Amber
Vacuum Desiccator
Cabinet
Polycarbonate
Hinged