missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ HDPE Jerry Can with Spigot

Catalog No. 11865135
Change view
Click to view available options
Capacity (Metric):
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11865135 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11865135 Supplier Bel-Art™ Supplier No. F118590025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Jerry can features a smooth turning spigot that opens and closes with half a turn for easy dispensing from a shelf or rack.

  • Feature smooth turning spigot that opens and closes with half a turn for easy dispensing from a shelf or rack
  • An extra hand grip on bottom gives positive control when pouring
  • Graduated in U.S. gallons and liters, they have extra headspace for expansion of contents
  • 20 cm (8 in.) long spout with a built-in air vent for smooth pouring
  • Spout retracts into a collar sealed by a leakproof screw cap when not in use
  • 5L can with 2.5 cm (1 in.) I.D. pouring spout and 10 to 20L with 1.9 cm (0.75 in.) I.D. spouts

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material HDPE
Closure Type Screw Cap
Color White
For Use With (Application) Liquid Storage
Graduations Liters, U.S. gal.
Includes Spigot
Material HDPE
Mouth Style Wide
Product Type Jerry Can
Shape Rectangular
Sterility Non-sterile
Show More Show Less