missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Flowmi™ Cell Strainers for 1000μL Pipette Tips

Catalog No. 14100150
Change view
Click to view available options
Mesh Size:
40 μm
70 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mesh Size
14100150 40 μm
03421228 70 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14100150 Supplier Bel-Art™ Supplier No. H136800040
Get Up to $500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis (patent pending). Available in two porosity sizes.

  • Provide fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis
  • Preserves volume when filtering small sample volumes
  • Removes cell aggregates and creates uniform single cell suspensions
  • Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
  • Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to tip; features a sliding cover for easy, one-handed use
  • Packaged in a re-sealable bag to maintain sterility when used in laminar flow hood or biohazard hood
  • Sterile, 40 μm or 70 μm porosity

Specifications

Mesh Material Polyethylene
Color White
For Use With (Application) Provide fast and efficient filtering of small volume samples (up to 1000μL) prior to FLOW or FACS™ analysis
Includes Compact tray
Product Type Cell Strainer
Sterility Sterile
For Use With (Equipment) Pipette Tips, 1000 μL, including Fisherbrand Sure-One, VWR, Axygen, Nichiryo, Eppendorf and more
Mesh Size 40 μm