missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Disposal Benchtop Carton for Glass

Catalog No. 03410590
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
03410590 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03410590 Supplier Bel-Art™ Supplier No. F246530002
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coated with 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids. Bel Art™ Disposal Benchtop Carton for Glass is easily closed for disposal.

Coated with 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids. Bel Art™ SP Scienceware™ Disposal Benchtop Carton for Glass is easily closed for disposal.

  • Holds 5.5 kg (12 lb)
  • Packaged flat to save space and shipping costs and quickly assembled
  • Lined with a 2 mil, clear polyethylene bag to store broken glass fragments and leaked liquids
  • Printed in four languages: English, German, French, Spanish
  • Safe, puncture-resistant receptacles for broken glassware
  • Does not qualify as a DOT container

Specifications

Capacity (English) 2.6 gal.
Color Red
Dimensions (L x W x H) 20 x 20 x 25 cm
Autoclavable Not Autoclavable
Includes Closure, Graphics in four languages (English, German, French, Spanish)
For Use With (Application) Glass
Material Corrugated Cardboard
Product Type Safety Container