missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ HDPE Dispensing Jug

Catalog No. 22260108
Change view
Click to view available options
Capacity (Metric):
20 L
Material:
HDPE
Product Type:
Dispensing Jug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Product Type
22260108 20 L HDPE Dispensing Jug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22260108 Supplier Bel-Art™ Supplier No. H118500000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to store and dispense common solutions, including janitorial cleaners, deionized and distilled water, or other aqueous liquids.

  • Storage capacity is 20 liters (5 gallons)
  • Gradations in gallons are molded into the side
  • Jug made from strong and lightweight HDPE
  • Dimensions: 25 x 38 cm (9-7/8 x 15 in.)

Specifications

Autoclavable Not Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material LDPE
Color White
For Use With (Application) To store and dispense common solutions, including janitorial cleaners, deionized / distilled water, or other aqueous liquids
Material HDPE
Mouth Style Wide
Product Type Dispensing Jug
Sterility Non-sterile