missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Cover for Glass Disposal Carton

Catalog No. 03421528
Change view
Click to view available options
Color:
White, Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03421528 White, Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03421528 Supplier Bel-Art™ Supplier No. F132040001
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Opens quickly and easily by pressing the built-in release tab with no batteries required

  • Store the disposable cardboard lid under the carton and, when carton is full, simply replace the reusable Disposal Carton Cover with the cardboard lid, and dispose of the carton in the usual fashion
  • Fits any disposal carton with a 12 x 12 in. opening including Glass Disposal Floor Standing Carton (Mfr. No. F24653-0001)
  • Features a smooth 10.1 x 10.1cm (4 x 4 in.) area for applying labels
  • Disposal Carton Cover provides a 20.3 x 24.1cm (8 x 9.5 in.) wider opening than the cardboard lids supplied, for safer, more sanitary deposit of wastes
  • The center panel is green to match the printing on the glass disposal cartons providing fast visual recognition of the waste stream
  • Wash the dishwasher-safe Disposal Carton Cover as needed, and it's ready for reuse
  • Autoclavable at 121°C (250°F)

Specifications

Color White, Green
Autoclavable Autoclavable
For Use With (Application) Provides safer sanitary deposit of wastes into the container
For Use With (Equipment) For disposal cartons with 12 x 12 in. opening, including glass disposal boxes
Product Type Carton Cover