Learn More
Bel-Art™ Connector Assortment
Catalog No. 22244558
Change view
Click to view available options
Shape:
Straight
Catalog No. Shape
22244558 Straight
1 options
Catalog No. 22244558 Supplier Bel-Art™ Supplier No. H195700000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Collection of polypropylene straight and straight-stepped tubing connectors
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7mm (0.125 to 0.5 in.).
  • Chemical-resistant polypropylene for use with wide range of liquids or gases
  • Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors
  • FDA-grade polypropylene for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Autoclavable Autoclavable
Material Polypropylene
Shape Straight
Color White
Product Type Tube Connector