Learn More
Bel-Art™ Cloning Cylinders
Polystyrene cloning cylinders facilitates the isolation of individual cell clones.
Supplier:  Bel-Art™ H378470000
 View more versions of this product
Catalog No. 0790710
Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left
Description
Description
Polystyrene cloning cylinders facilitates the isolation of individual cell clones.
  • Tapered
  • Good sealing surface
  • Polystyrene
  • Sterile
Order Info
Individual sizes supplied five bags of 10 cylinders each; Variety Pack contains four bags with 10 of each size
Specifications
Chemical Identifiers
Sterile
Polystyrene
Cloning Cylinder
Specifications
4.7 mm, 6.4 mm, 9.5 mm
4.7, 6.4, 9.5 mm
Polystyrene
Cloning Cylinder
Small, Medium, Large (Variety)
8 mm, 11 mm
Sterile
0.185 in., 0.251 in., 0.37 in.
Isolation of individual cell clones
Product Suggestions
Product Suggestions