missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Clavies™ Biohazard Bag Holder Cover

Catalog No. 03410600
Change view
Click to view available options
Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03410600 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03410600 Supplier Bel-Art™ Supplier No. F131920102
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Opens quickly and easily by pressing the built-in release tab — no batteries required

  • Cover installs in seconds to improve sanitation of biohazard waste containers
  • Dishwasher and autoclave safe: autoclavable at 121°C (250°F)
  • Fits both the medium and large sizes of the Clavies Biohazard Bag Holders (Mfr. Nos. 13192-0002 and 13192-0003)
  • Features a smooth 4 x 4 in. (10.1 x 10.1cm) area for applying labels
  • Center panel of the lid opens to an upright position and reveals a large 8 x 9.5 in. (20.3 x 24.1cm) opening
  • Orange

Specifications

Color Orange
Autoclavable Autoclavable
For Use With (Application) Improves the sanitation of biohazardous waste containers
For Use With (Equipment) Clavies™ biohazard bag holders
Product Type Biohazard Bag Holder Cover