missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves

Catalog No. p-144199
Change view
Click to view available options
Length (English) Cuff:
5 in.
11 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Cuff
11394299 5 in.
11394298 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11394299 Supplier Bel-Art™ Supplier No. H13201 0000
Only null left

Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).

  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 232°C
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer

Specifications

Certifications/Compliance CE
Accelerator Free Yes
Applications Laboratory, Industrial
Coating Material Uncoated
Color Orange
Latex Free Yes
Material Terrycloth
Product Type Autoclave Gloves
Size Universal
Style Full-hand Mitt
Temperature (English) Max. 450°F
Temperature (Metric) Max 232°C
For Use With (Application) Handling Hot Objects
Glove Cuff Style Gauntlet
Series H13201
Length (English) 13 in.
Length (English) Cuff 5 in.
Length (Metric) 33 cm
Length (Metric) Cuff 13 cm
Show More Show Less