Learn More
Bel-Art™ Churn Sample Splitter
Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis
Supplier:  Bel-Art™ F378050008
 View more versions of this product
Catalog No. 0333812B
Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left
Description
Description
  • Polyethylene sample splitter consists of thick walled cylindrical vessel with disc churn
  • Center handle extends through cover and locating notch keeps it oriented in one direction
  • Small, polypropylene spring-loaded spigot drains subsamples into bottles for delivery to water quality laboratory
  • Heavy duty swing type handles supports full or empty churn
  • Built-in full open lever stop on spigot ensures reproducible dispensing of subsamples
  • Water sediment samples are poured into sample splitter where they are composited into one representative cross section sample of stream flow
  • Lid, disc churn and spigot are replaceable
  • Intended to meet USGS water sampling protocol
Specifications
Chemical Identifiers
Churn Sample Splitter
Polyethylene
Not Autoclavable
Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis
Specifications
Churn Sample Splitter
Not Autoclavable
2.1 gal.
Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis
28 cm
Polyethylene
Thick walled cylindrical vessel with disc churn, small polypropylene spring-loaded spigot, built-in full open lever, heavy duty swing type handle
250 mL
11.02 in.
11 in.
Product Suggestions
Product Suggestions