missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Captor™ Pen Holder

Catalog No. 0181613
Change view
Click to view available options
Quantity:
3 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0181613 3 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0181613 Supplier Bel-Art™ Supplier No. F133730000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cap and uncap pen with one hand

  • Avoid fumbling with caps and risking contamination of samples
  • Fits Sharpie™ markers or similar pens with a pocket clip
  • Attaches with press-fit Dual Lock™ reclosable fastener to any flat surface, including incubator, freezer, centrifuge, fume hood or benchtop
  • Black polycarbonate
  • 1L × 1W × 2 in.H (2.5 × 2.5 × 5.1cm)

Specifications

Tip Style Felt Tip
Product Type Pen Holder
Color Black
Material Polycarbonate
For Use With (Application) Fits Sharpie™ markers or similar pens with a pocket clip
For Use With (Equipment) Attaches with press-fit Dual Lock reclosable fastener
Quantity 3 Pk.
Height (English) 2 in.
Length (English) 1 in.
Width (English) 1 in.