missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Sharp Object Safety Pouch

Catalog No. 0181517
Change view
Click to view available options
Capacity (Metric):
1100 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0181517 1100 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0181517 Supplier Bel-Art™ Supplier No. H132340000
Get Up to $500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers protection against cuts by contaminated sharp objects during work in labs, clinics, and medical offices

  • Pouch remains open in an oval shape by folding the bottom flaps
  • After filling, the top flaps close and lock in place for safe disposal
  • Solid paperboard pouch is 10 mil thick resists puncture by sharps
  • Printed with bold biohazard symbol
  • Measures 14 x 33 cm (5.5 x 13 in.) flat

Order Info

Safety pouch stand (22-197-061 ) sold separately.

Specifications

Capacity (Metric) 1100 cc
Capacity (English) 0.29 gal.
Color White
Autoclavable Not Autoclavable
For Use With (Application) For collection and disposal of sharp objects
Material Corrugated Cardboard
Product Type Sharps Container