missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Incinerator Disposal Carton

Catalog No. 22279178
Change view
Click to view available options
Capacity (English):
14.4 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22279178 14.4 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22279178 Supplier Bel-Art™ Supplier No. F132050001

Includes polyethylene liner. Bel Art™ Biohazard Incinerator Disposal Cartons are clearly marked for proper handling and processing.

  • Corrugated cardboard carton features fitted lid with flap
  • Lined with clear 1.5 mil polyethylene biohazard bag that resists tears, punctures
  • Bright biohazard markings for immediate and continuous identification
  • Instructions in English, Spanish, French and German
  • Use with Bel-Art reusable cover for biohazard carton (Mfr. No. F132040000)
  • Dimensions 12 x 12 x 27 in.

Specifications

Capacity (English) 14.4 gal.
Color Red, White
Dimensions (L x W x H) 12 x 12 x 27 in. (30.5 x 30.5 x 68.6 cm)
Height (Metric) 68.6 cm
Length (Metric) 30.5 cm
Width (Metric) 30.5 cm
Autoclavable Not Autoclavable
Liner Material Polyethylene
For Use With (Application) Proper handling and disposal of biohazard waste
Width (English) 12 in.
Length (English) 12 in.
Thickness 1.5 mm
Height (English) 27 in.
Material Cardboard
Product Type Biohazard Incinerator Carton Container
Show More Show Less