missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Bacterial Cell Spreaders
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
$87.59 - $92.38
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Material | Stainless Steel |
| Product Type | Bacterial Cell Spreader |
| Sterility | Sterile |
| Length (English) | 12 in. |
| Catalog Number | Mfr. No. | Size (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size (Metric) | Price | Quantity & Availability | |||||
|
087692A
|
Bel-Art™
F377360006 |
16 mm Spreader Bar |
Each for $92.38
|
|
|||||
|
087692B
|
Bel-Art™
F37736 0009 |
25 mm Spreader Bar |
Each for $87.59
|
|
|||||
Description
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
- Autoclavable and may be flame sterilized
- Features triangular end with smooth rounded surfaces throughout
- Smaller 16mm width bar suitable for use with 6cm plates
- Larger 25mm width bar suitable for use with 9cm plates
- Use with Inoculating Turntable (Mfr. No.F40913-0000)
- Both sizes 30cm long
Specifications
| Autoclavable | |
| Bacterial Cell Spreader | |
| 12 in. | |
| Easily perform inoculation of petri dishes |
| Stainless Steel | |
| Sterile | |
| 30.2 cm |