missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Automatic Self-Zeroing Burets
Feature easy-to-read blue graduations against a white background
Supplier: Bel-Art™ H379180110
Description
- The delivery end is fitted into a plastic push button stopcock which permits drop by drop delivery
- Small screw clamp adjusts drop speed as needed for titration
- Polyethylene filling tube extends from the top of the burette into the bottle reservoir
- Graduation accuracy is Class B
- Glass burette has 1-10 divisions
- Bottle, screw cap and round base are polyethylene plastic
Chemical Identifiers
| Class B | |
| Glass | |
| Bottle, Round Base, Screw Cap | |
| Automatic Buret |
| Screw Cap | |
| 0.1 mL | |
| Blue | |
| Class B |
Specifications
| Class B | |
| Screw Cap | |
| 15 mL | |
| Polyethylene 500 mL Reservoir | |
| 0.1 mL | |
| Bottle, Round Base, Screw Cap | |
| Yes | |
| +/- 0.015 mL | |
| 500 mL | |
| Class B |
| Not Autoclavable | |
| White | |
| 52 cm | |
| Glass | |
| Reagents | |
| Blue | |
| Plastic | |
| No | |
| Automatic Buret |