missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Air Operated Turbine Magnetic Stirrer
Description
Includes
Tubing is supplied to connect to an air supply.
- Uses only 3 to 8 psi (21 to 55 kPa) to drive the magnetic stirrer
- Excellent for use where potentially explosive solvent vapor are present
- Can be used with magnetic stirring bars up to 2.5 cm (1 in.) in length
- Eliminates sparking hazard from electrical sources
- Made from high impact polystyrene to ensure long life and easy clean-up in case of spills
Specifications
Specifications
| Speed | 0 to 2000 rpm |
| Stirring Positions | 1 |
| Top Plate Material | Polystyrene |
| Capacity | 1 L |
| Display | None |
| Shape | Round |
| Voltage | 0/1 V |
| Dimensions (L x W) | 4 x 0.75 in. (102 x 19 mm) |
| Length (English) | 4 in. |
| Length (Metric) | 102 mm |
| Show More |