missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Acid/Solvent Bottle Carriers

Catalog No. p-51677
Change view
Click to view available options
Holds:
Bottles (2.5 L)
Bottles (4 L)
Material:
Polyethylene
Polypropylene
Height (Metric):
35 cm
37 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Material Height (Metric)
22260395 Bottles (4 L) Polyethylene 37 cm
22260397 Bottles (4 L) Polypropylene 37 cm
22260394 Bottles (2.5 L) Polypropylene 35 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22260395 Supplier Bel Art™ Supplier No. F169580000
Only null left

Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles

  • Offer shatter resistant protection for glass bottles
  • Cone style lid has a 0.625 in. (16 mm) opening in top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles

Specifications

Autoclavable Not Autoclavable
Color White, Blue
Holds Bottles (4 L)
Material Polyethylene
Positions 1
Shape Cylindrical
Stackable No
Sterility Non-sterile
Length (Metric) 370 mm
Height (English) 14.37 in.
Includes Rack Only
Product Type Bottle Carrier
Diameter (Metric) Inner 200 mm
Shelves None
Height (Metric) 37 cm
Array 1 x 1
Width (Metric) 200 mm
Show More Show Less