missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ 28-Piece Fittings Assortment Kit

Catalog No. 1371723
Change view
Click to view available options
For Use With:
0.19 to 0.5 in. Tubing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
1371723 0.19 to 0.5 in. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1371723 Supplier Bel-Art™ Supplier No. F196260000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compatible with tubing from 3.2 to 12.7mm. Bel Art™ 28-Piece Fittings Assortment Kit are for the most common lab connections.

  • Molded of autoclavable FDA grade polypropylene except quick disconnect fittings which are polyethylene and not autoclavable or FDA grade
  • Kit contains: 6 Straight Tubing Connectors, 7 Stepped Tubing Connectors, 5 Wye (Y) Tubing Connectors, 3 “T” shaped Tubing Connectors, 5 Quick Disconnects and 2 flow control Plastic Tubing Clamps
  • Plastic organizer box has room for adding other fittings and parts

Specifications

Autoclavable Autoclavable
Material Polypropylene
For Use With 0.19 to 0.5 in. Tubing
Color Natural
Product Type Fittings Kit