missing translation for 'onlineSavingsMsg'
Learn More

Behenic Acid 80.0+%, TCI America™

Catalog No. D096325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096325G 25 g
D0963500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D096325G Supplier TCI America Supplier No. D096325G

Chemical Identifiers

CAS 112-85-6
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.592
MDL Number MFCD00002807
InChI Key UKMSUNONTOPOIO-UHFFFAOYSA-N
Synonym behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
PubChem CID 8215
ChEBI CHEBI:28941
IUPAC Name docosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 79°C
Color White
Boiling Point 306°C
Quantity 25 g
Formula Weight 340.59
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Behenic Acid
EINECSNumber (2)-0608
TSCA Yes