missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Behenic Acid | |
| 80°C | |
| 306°C | |
| MFCD00002807 | |
| behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)O | |
| 340.592 | |
| CHEBI:28941 | |
| ≥95.0% (GC) |
| 112-85-6 | |
| White-Yellow | |
| C22H44O2 | |
| 25 g | |
| UKMSUNONTOPOIO-UHFFFAOYSA-N | |
| docosanoic acid | |
| 8215 | |
| 340.59 | |
| Crystalline Lumps |
Chemical Identifiers
| 112-85-6 | |
| 340.592 | |
| UKMSUNONTOPOIO-UHFFFAOYSA-N | |
| 8215 | |
| docosanoic acid |
| C22H44O2 | |
| MFCD00002807 | |
| behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure | |
| CHEBI:28941 | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)O |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : Yes