Learn More
Waters Limited ACQUITY - XBridge BEH HILIC Method Transfer Kit, 130, 1.7 - 3.5 m, 2.1 - 4.6 mm X 100 - 150mm, 2/pk
Catalog No. 50791277
Encompass
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms. Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power. Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
Catalog No. 50791277 Supplier Waters Limited Supplier No. 186004973
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms. Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power. Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.