Learn More
Lonza Walkersville Inc BEGM Bronchial Epithelial Cell Growth Medium BulletKit
Supplier:  Lonza Walkersville Inc CC3170
Encompass
BEGM Bronchial Epithelial Cell Growth Medium is specially designed to support the growth of human primary bronchial epithelial cells. BEGM BulletKit is serum-free and has been optimized to grow cells from normal and diseased tissues.Published research shows BEGM BulletKit has expanded applications. Our customers have used BEGM BulletKit to culture well-established cell lines such as BEAS-2B, home-grown bronchial epithelial cells and supporting co-culture models with breast cancer cell lines such as MCF-7 and MDA-MB-231. Refer to the published literature in the 'Data' tab.BEGM is an optimized and readily available media kit providing researchers the confidence and the convenience of a system that works to support their cells. This is especially important for researchers who are utilizing cell lines as a stepping stone to primary cell culture.
Catalog No. NC9202780
Only null left