Learn More
Carson™ Beginner 100X to 1000X Compound Student Microscope DFS Item
Carson™ Beginner 100X to 1000X Compound Student Microscope features a custom retrofitted universal smartphone digiscoping adapter for use with the built-in smartphone camera application to record video and take images through the eyepiece of the microscope.
Supplier:  Carson™ MS100SP
Catalog No. S81146
Only null left
Explore available promotions
Description
Description
  • Digiscoping adapter connector is retrofitted to fit over the included microscope eyepiece.
  • 100X to 1000X table-top microscope is an intermediate level biological microscope that is ideal for a high school/university student, hobbyist or technicians.
  • Metal body
  • Monocular head that sits at a 45° angle for comfortable viewing.
  • Monocular head rotates to 360° for easy sharing of the specimen view.
  • Both coarse and fine focus knobs to help the user to achieve a sharp focus.
  • LED illumination provides a bright white colored light and also has an intensity control to set the optimal lighting conditions.
  • Filter holder with an included blue color filter.
  • Included AC power adaptor allows for use internationally without the need for a voltage transformer.
  • Compatible with dual-camera phones.
  • Horizontal phone alignment is done using a variable pitch screw for fine control that can be moved quickly between an iPhone (corner camera) and Android (center camera) positions.
  • Alignment features will not interfere with any smartphone side buttons.
  • One year limited warranty
  • Fits all phones
Specifications
Specifications
10X, 40X, 100X
MS-100SP
Viewing samples under high magnifications
Mechanical Stage and Universal Smartphone Digiscoping Adapter
Product Suggestions
Product Suggestions