missing translation for 'onlineSavingsMsg'
Learn More

High-Tech Conversions Bee-Safe™ Super Non-Slip Shoe Covers

Catalog No. 17444139
Change view
Click to view available options
Closure Type:
Elastic Ankle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
17444139 Elastic Ankle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17444139 Supplier High-Tech Conversions Supplier No. GAHSCSXL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bee-Safe™ Super Non-Slip Shoe Covers provide a super sticky base that is designed to reduce slipping and sliding. These sticky polyethylene, nonwoven coated shoes covers are ideal for use in areas that need to be protected from outside contaminants.

  • Light liquid splash protection/water resistance
  • Soft non-woven polypropylene for greater comfort
  • Latex-free elastic edges to minimize allergic reactions
  • Machine made to ensure consistency throughout production

Specifications

Size X-Large
Material Polyethylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
For Use With (Application) Laboratory, Medical Devices, Aerospace, Electronics, Food, Industrial
Packaging Quantity 300 Pk.
Packaging Type Bag
Product Type Controlled Environments Shoe Covers
Show More Show Less