missing translation for 'onlineSavingsMsg'
Learn More

High-Tech Conversions Bee-Safe™ Non-Skid Cleanroom Shoe Covers

Catalog No. 17444039
Change view
Click to view available options
Size:
X-Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17444039 X-Large
17444040 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17444039 Supplier High-Tech Conversions Supplier No. GAHSPNSXL

Non-skid, non-woven shoe covers are ideal for use in laboratories and manufacturing environments.

  • Low-linting nonwoven fabric helps filter particulates for contamination control
  • Non-skid soles for enhanced traction
  • Elastic band gives secure yet comfortable fit around the shoes
  • Ideal for lab and light industrial use
  • For use in laboratories and manufacturing environments

Specifications

Size X-Large
Material Spunbond Polypropylene
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
For Use With (Application) Laboratories, Manufacturing
Packaging Quantity 300/Cs.
Length (English) 17 in.
Length (Metric) 43 cm
Height (English) 7 in.
Height (Metric) 18 cm
Product Type Controlled Environments Shoe Covers
Show More Show Less