Learn More
High-Tech Conversions Bee-Safe™ Open Face Cleanroom Hood
Catalog No. 17444043
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17444043 Blue
17444042 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17444043 Supplier High-Tech Conversions Supplier No. GAHHOODBLL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Open face cleanroom hood is lightweight with particulate filtration.
  • Polypropylene cleanroom hood is ideal for a variety of applications including light industrial, lab and cleanroom
  • Provides complete head coverage and is made of lightweight, low-linting, and breathable polypropylene material
  • Latex-free
Specifications
Product Type Open Face
Material Spunbond Polypropylene
Color Blue
Closure Type Elastic
Model GAH-HOOD
Size (English) OSFM
Sterility Non-sterile
Quantity 100/Bag, 10 Bags/Cs.