missing translation for 'onlineSavingsMsg'
Learn More

Beclometasone Dipropionate 98.0+%, TCI America™

Catalog No. p-7123556
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44641G 1 g
B44645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44641G Supplier TCI America Supplier No. B44641G
Only null left

Chemical Identifiers

CAS 9-8-5534
Molecular Formula C28H37ClO7
Molecular Weight (g/mol) 521.05
MDL Number MFCD00135613
InChI Key KUVIULQEHSCUHY-XYWKZLDCSA-N
Synonym beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
PubChem CID 134129500
IUPAC Name [2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 1 g
Formula Weight 521.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Beclometasone Dipropionate
RTECSNumber TU3805000
TSCA No