missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclometasone-17-Propionate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 5534-18-9 |
| Flash Point | Not Applicable |
| Molecular Formula | C25H33ClO6 |
| Linear Formula | C25H33ClO6 |
| Quantity | 25 mg |
| Formula Weight | 464.98 |
| Grade | British Pharmacopoeia (BP) Reference Standard |