missing translation for 'onlineSavingsMsg'
Learn More

HemoCue America Beckman Coulter™ ICON™ 25 hCG Test Kit

Catalog No. 22247114
Encompass
Change view
Click to view available options
Clia Complexity:
Waived, Non-waived
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22247114 Waived, Non-waived
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22247114 Supplier HemoCue America Supplier No. 43025A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fast, accurate results for pregnancy testing right in the office

  • CLIA Complexity: Waived (urine), Non-waived (serum)
  • Sensitivity: 25 mIU/mL
  • Specificity: >99%
  • Simple, one-step process for reliability
  • Requires just three drops of specimen
  • Three-minute result with urine; five-minute result with serum
  • Two distinct red/purple lines indicate positive results

Specifications

Content And Storage Room Temperature
Clia Complexity Waived, Non-waived
Sample Type Serum, Urine
Sensitivity As low as 25 mIU/mL
Type hCG Test Kit
Sample Size 3 Drops
Specificity >99%
Test Time 3 min. for urine, 5 min. for serum