Learn More
HemoCue America Beckman Coulter™ Hemoccult™ SENSA™ Fecal Occult Blood Slide Test System: Supplies
Catalog No. SK64115
Encompass
Change view
Click to view available options
Quantity:
20 Pack
Type:
Developer
Catalog No. Quantity Type
SK64115 20 Pack Developer
1 options
Catalog No. SK64115 Supplier HemoCue America Supplier No. 64115
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For detection of fecal occult blood
Supplies for SENSA Fecal Occult Blood Slide Test System.
Specifications
CPT Code 82270, G0107
Clia Complexity Waived
Type Developer
For Use With (Application) Detection of fecal occult Blood
Includes Developer (15mL)
Quantity 20 Pack