missing translation for 'onlineSavingsMsg'
Learn More

HemoCue America Beckman Coulter™ Hemoccult™ Fecal Occult Blood Slide Test System

Several packaging configurations are available for different testing needs

Supplier:  HemoCue America 61130A

Encompass

 View more versions of this product

Catalog No. SK61130


Only null left

Description

Description

  • CLIA Complexity: Waived
  • CPT Codes: 82270, G0107
  • A convenient, qualitative procedure that uses a guaiac paper slide test to detect fecal occult blood
  • On-slide performance monitor enables function and stability of reagents to be tested each time a slide is developed
  • Test aids in reducing colorectal cancer with annual screening
  • Results in 60 seconds
  • Positive results appear as a blue development on the edge of the fecal smear
  • Hemoccult II™ Slides feature cards of three tests designed to allow patients to collect three serial specimens at home
  • Dispensapak™ and Dispensapak Plus™ Packs are designed for at-home use by patient
  • Accessories available; see cat. no. SK63202

Specifications

Chemical Identifiers

82270, G0107
Waived
60 sec.

Specifications

82270, G0107
Dispensapak Plus Kit
40 Pack
Waived
6 x 15mL Bottles of developer and 40 patient kits (1 triple slide, 3 flushable sample collection tissues, 3 applicator, diet and test instructions and barrier mailing envelope)
60 sec.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions