Learn More
Thermo Scientific™ Beaker Racks for Solaris™ Open Air Shakers & Temperature Controlled Shakers, and MaxQ™ 6000 Shakers
Mix and match Thermo Scientific™ Solaris shaker accessories to suit individual needs in the laboratory. Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers.
Supplier: Thermo Scientific™ 75004119
Description
Mix and match Thermo Scientific™ Solaris shaker accessories to suit individual needs in the laboratory. Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers.
Mix and match Thermo Scientific™ Solaris shaker accessories to suit individual needs in the laboratory
- Racks can hold beakers from 30 - 600 mL
- Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers, MaxQ6000 Shakers
Chemical Identifiers
| Beaker Rack |
Specifications
| 9 Beakers | |
| 11 x 14 in. or 12 x 14 in. | |
| Rack for 250 mL beaker |
| Beaker Rack | |
| 351 x 286 x 58 mm | |
| Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers |