missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Omni International Bead Ruptor 96 96-well 2D Barcoded Storage Tube Adapter for Micronic and Fluidx Tubes

The Bead Ruptor 96-well 2D Barcoded Storage Tube Adapter for Micronic and Fluidx Tubes is a 96 place rack accessory.
Supplier: Omni International 27107
Description
- Includes a pair of two stainless steel 2D barcoded storage tube adapters
- Each pair will hold one rack of 96 x 1.1 mL or 1.4 mL 2D barcoded storage tubes
- Short Compatible: FluidX, Micronic MP32120, FluidX 68-1000-00, Thermo Fisher Scientific Matrix 3790
Specifications
| Tube Adapter | |
| Autoclavable | |
| Bead Ruptor 96 | |
| 1 rack of 96 x 1.1 mL or 1.4 mL 2D Barcoded Storage Tubes |