Learn More
BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Hemogard™ Closure
Use for serum determinations in chemistry, serology and immunohematology
$51.07
Specifications
Additive Clot Activator, Silicone Coated
Material Plastic
Closure Type Hemogard
Closure Color Red
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer For Use With (Application) Price Quantity & Availability  
0268794
View Documents Promotion Details
BD
367814
5 mL 13 x 100 mm Blood Bank Use Not Recommended
Pack of 100 Each for $51.07
Case of 10 Pack for $510.70
Only null left
0268394
View Documents Promotion Details
BD
367815
6 mL 13 x 100 mm Blood Bank Use Not Recommended
Pack of 100 Each for $51.07
Case of 10 Pack for $510.70
Only null left
02685111
View Documents Promotion Details
BD
367812
4 mL 13 x 75 mm Blood Bank Use Not Recommended
Pack of 100 Each for $51.07
Case of 10 Pack for $510.70
Only null left
Description
Description
  • Sterile vacuum blood collection tubes for serum determination
  • Spray-coated silica interior
  • With Hemogard™ tube closure, which prevents human contact with blood on the stopper or tube rim
Evacuated Tubes Simplify Blood Collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Plastic Evacuated Tubes
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Hemogard™ Stoppers
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
Specifications
Specifications
Clot Activator, Silicone Coated
Plastic
Hemogard
Red