missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Rapid Serum Tube

Catalog No. 23021640
Change view
Click to view available options
Capacity (Metric):
5 mL
Closure Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
23021640 5 mL Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23021640 Supplier BD Supplier No. 368774
Only null left

Ready for analysis in only 8 minutes

  • With Hemogard™ closure
  • Reduces contact with blood on stopper and blood splatter during de-capping
  • Compatible with major laboratory automation systems
  • V-Notch™ Label: Helps align secondary label
  • Thrombin based clot activator: Promotes rapid clotting
  • DEHP-free gel seperator: Extended analyte stability
  • 3 minute spin at 4000 x g (RCF)

Specifications

Product Type Rapid Serum Tube
Additive Thrombine
Material Plastic
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 5 mL
Closure Color Orange
Tube Size 13 x 100 mm
For Use With (Application) Serum Determinations, STAT Testing
Label Type Paper
Length (Metric) 100 mm
Diameter (Metric) Outer 13 mm
Show More Show Less