Learn More
BD Vacutainer™ Pronto™ Quick Release Needle Holder
Catalog No. 0266511
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
0266511 Plastic
1 options
Catalog No. 0266511 Supplier BD Supplier No. 368872
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to prevent over-tightening when needle is secured
  • Reusable plastic holder (100 estimated uses per holder), designed for use with all BD Vacutainer single and multiple sample needles, blood collection sets and Luer adapter
  • Designed to prevent overtightening when needle is secured and to facilitate a smooth and trouble-free release
  • Requires no assembly and can be used with any sharps container
Specifications
For Use With (Equipment) BD Vacutainer single and multiple sample needles, blood collection sets and luer adapter
For Use With (Application) Prevent over-tightening when needle is secured
Product Type Quick Release Needle Holder
Material Plastic