missing translation for 'onlineSavingsMsg'
Learn More

BD PAXgene™ Blood RNA Tube

Catalog No. 2302101
Change view
Click to view available options
Capacity (Metric):
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
2302101 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2302101 Supplier BD Supplier No. 762165
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers a convenient and closed system for the collection, storage and transportation of whole blood that requires intracellular RNA stabilization. BD Vacutainer™ PAXgene™ Blood RNA Tube is for in vitro diagnostic use and is CE-marked.

The collection of whole blood is the first step in many molecular assays used to study intracellular RNA. A major challenge in this type of testing is the instability of intracellular RNA, which rapidly degrades within hours after blood collection. Furthermore, certain species of RNA, through the process of gene induction, increase in vitro after blood collection. Both in vitro RNA degradation and gene induction can lead to an under- or over-estimation of in vivo relative gene transcript number.

The PAXgene™ Blood RNA Tube contains an additive that stabilizes the in vivo gene transcription profile by reducing in vitro RNA degradation and minimizing gene induction. When used in conjunction with the PAXgene™ Blood RNA Kit, the Blood RNA Tube provides for accurate detection and quantitation of gene transcripts. Store unused PAXgene Blood RNA Tubes at 18–25°C. (Limited excursion temperatures up to 40°C are permitted.)

Specifications

Color Clear
Product Type Cell Preparation Tube
Additive PAXgene Blood RNA Preservative Solution, 6.9mL
Material PET
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 2.5 mL
Includes 100/case
For Use With (Application) Molecular Diagnostic Testing
Label Type Paper
Length (Metric) 100 mm
Diameter (Metric) Outer 16 mm
Certifications/Compliance CE marked
Show More Show Less