Learn More
BD Vacutainer™ Glass Mononuclear Cell Preparation (CPT) Tubes
Evacuated tube intended for the collection of whole blood and the separation of mononuclear cells
$1,958.16 - $1,977.47
Specifications
Closure Type Traditional Cap
Capacity (Metric) 8 mL
View More Specs
Products 2
Catalog Number Mfr. No. Closure Color Additive Price Quantity & Availability  
Catalog Number Mfr. No. Closure Color Additive Price Quantity & Availability  
02685125
View Documents Promotion Details
BD
362761
Light Blue/Black Sodium Citrate
Case of 60 Each for $1,958.16
Only null left
1495951D
View Documents Promotion Details
BD
362753
Red/Green Sodium Heparin and Polymer Gel
Case of 60 Each for $1,977.47
Only null left
Description
Description
BD Vacutainer™ Mononuclear Cell Preparation Tube (CPT™)
  • The one-step, closed-system tube for blood collection, mononuclear cell separation and transportation
  • Contributes to increased lab productivity by offering convenience, safety and reproducibility
  • CPT Cell Preparation tubes are an evacuated blood collection tube system containing either 0.1 M sodium citrate anticoagulant or sodium heparin anticoagulant
  • Tubes also contain blood separation media composed of a thixotropic polyester gel and a FICOLL™ Hypaque™ solution
Specifications
Specifications
Traditional Cap
8 mL