missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Collection Tube

Catalog No. 0268345
Change view
Click to view available options
Additive:
Sodium Fluoride, Sodium EDTA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
0268345 Sodium Fluoride, Sodium EDTA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0268345 Supplier BD Supplier No. 365992

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for ease-of-use and ensuring that a quality capillary blood sample is collected

  • Wider-diameter BD Microtainer Tube
  • BD Microgard™ closure for safety
  • Color coded to match the array of BD Vacutainer evacuated blood collection tubes
  • With sodium fluoride and sodium-EDTA added
  • Volume: 400 to 600μl
  • Color: gray

Specifications

Product Type Capillary Tube
Additive Sodium Fluoride, Sodium EDTA
Sterility Sterile
Closure Included Yes
Closure Color Gray
For Use With (Application) Blood glucose testing