Learn More
BD Syringe with Luer-Lok Tips (Without Needle)
Siliconized and assembled with scale and tip shield
Supplier:  BD 301033
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-823-16K
Il en reste null
Découvrez plus d'offres spéciales
Avertissement:
NOTICE: The printing has changed on BD 60 mL syringes to only be graduated to 50 mL now. The syringe itself has not changed. Syringes graduated to 60 mL are no longer available.
Description
Description
Siliconized and assembled with scale and tip shield
  • Single-use with plastic barrels, plungers and tips
  • Nonsterile syringes may by irradiated, autoclaved, or gas sterilized out of polybag
Spécifications
Spécifications
Plastic
1 mL
30 mL
Non-sterile
Syringe
No Needle Included
Luer Lock
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS