Learn More
BD SafetyGlide™ Needles
Catalog No. B305917
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Length (English) Needle:
0.62 in.
1 in.
1.5 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Needle
B305917 21 G 1.5 in.
14826106 25 G 1 in.
1482910 25 G 0.62 in.
1482911 23 G 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B305917 Supplier BD Supplier No. 305917
Needles feature patented Activation-Assist™ Technology for fast and easy, single-handed needle-tip shielding immediately after injection.
  • This technology provides versatility and minimal change in technique
Specifications
Needle Gauge 21 G
For Use With (Application) General and hypodermic use
Includes Shielding mechanism
Latex Free Yes
Length (English) Needle 1.5 in.
Product Type Shielding Hyperdermic Needle
Needle Point Style Regular Bevel
Sterility Sterile