missing translation for 'onlineSavingsMsg'
Learn More

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 (LP80)

Catalog No. B215278
Change view
Click to view available options
Quantity:
100/Pk.
Media Type:
TSA w/Lecithin, Polysorbate 80
Packaging Type:
Single Wrapping, Aseptic Fill
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type Packaging Type
B215278 100/Pk. TSA w/Lecithin, Polysorbate 80 Single Wrapping, Aseptic Fill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B215278 Supplier BD Supplier No. 215278
Only null left

The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.

The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:

  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type TSA w/Lecithin, Polysorbate 80
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Packaging Type Single Wrapping, Aseptic Fill
Quantity 100/Pk.