missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD RODAC™ SNAP LID D/E Neutralizing Agar
Description
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
- Withstands an accidental drop from the height of a bench-top workspace
- Ensures the integrity of the sample
- Prevents the dish from opening unintentionally
- Protects media from accidental cross contamination
- Improves stability during stacking and storage
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Purple |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | D/E neutralizing Agar |
| Sterility | Sterile |
| Includes | Rodac™ snap lid |
| Packaging Type | Sterile Double Wrapping Plate |
| Quantity | 100/Pk. |
| Weight (Metric) | 2720 g |