missing translation for 'onlineSavingsMsg'
Learn More

BD DMACA Indole Reagent

Numéro de catalogue. B4361187
Modifier l'affichage
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B4361187 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. B4361187 Fournisseur BD Code fournisseur 261187
Il en reste null
Edge

Identify microorganisms without mixing or measuring reagents

To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

  • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
  • Strict quality controls assure uniformity and accuracy

Spécifications

Type DMACA Indole Reagent
Quantity 50 Pack
Certifications/Compliance CE
Format Dropper
For Use With (Application) Used for the rapid identification of bacteria containing tryptophanase enzyme systems
Storage Requirements 15° to 30°C
Stain Type DMACA Indole